3-Nitrofluoranthene-[D9] 50 µg/ml (98% D), CIL
Catalog # PROOCILDLM47111.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:892-21-7
- Description:3-Nitrofluoranthene-[D9] 50 µg/ml (98% D)
- Formula:C₁₆D₉NO₂
- Description:3-Nitrofluoranthene-[D9]