
4-(Dimethylamino)cinnamaldehyde for synthesis, Sigma-Aldrich®
Catalog # 8.22034.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:135 - 138 °C
- Molar Mass:175.22 g/mol
Specification Test Results
About this item
:
:175.23 g/mol
:M-Clarity™ quality level = MQ200
EINECS:228-267-0
:GHS07
Storage Temperature:Ambient
CAS Number:6203-18-5
Formula:C₁₁H₁₃NO
MDL Number:MFCD00007002