
4-(Dimethylamino)cinnamaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6203-18-5
:
Formula:C₁₁H₁₃NO
MDL Number:MFCD00007002
:175.23 g/mol
Melting Pt:137…144 °C
EINECS:228-267-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together