
4-(Dimethylamino)cinnamaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6203-18-5
:
Formula:C₁₁H₁₃NO
MDL Number:MFCD00007002
:175.23 g/mol
Melting Pt:137…142 °C
EINECS:228-267-0
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Yellow to orange Powder
- Infrared spectrum:Conforms
- Melting point:137 to 142 °C
- Titration with HClO4:≥97.5 %
- Stabilizer:0.1 % Hydroquinone
Specification Test Results
Frequently Bought Together