N-Ethylpyridine-4-methylamine, Sigma-Aldrich®
Catalog # SIAL367095-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:136.20 g/mol
:M-Clarity™ quality level = MQ100
EINECS:251-506-5
CAS Number:33403-97-3
Formula:C₈H₁₂N₂
MDL Number:MFCD00023632