6-Cyano-2-naphthol, Sigma-Aldrich®
Catalog # SIAL530263-25G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:25 g
- Sekvence:3308
Specifications
About this item
:
:169.1794 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:52927-22-7
Formula:C₁₁H₇NO
MDL Number:MFCD01863480