Diethyl phosphoramidate, Sigma-Aldrich®
Catalog # SIAL363596-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:153.1167 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:1068-21-9
Formula:C₄H₁₂NO₃P
MDL Number:MFCD00015676