5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:102185-33-1
:
Formula:C₈H₄BrClNNa₂O₄P
MDL Number:MFCD00036757
:370.43 g/mol
:M-Clarity™ quality level = MQ200
Frequently Bought Together