5-Bromo-4-chloro-3-indolyl phosphate disodium salt hydrate (BCIP-Na hydrate) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:205926-98-3
:102185-33-1
Formula:C₈H₄BrClNNa₂O₄P·xH₂O
MDL Number:MFCD00150468
:370.43 g/mol (anhy)
Melting Pt:300 °C
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to light beige to light bleu Powder
- Infrared spectrum:Conforms
- Water:5 to 9 % (K.F.)
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together