
Diethyl benzylphosphonate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1080-32-6
:
Formula:C₁₁H₁₇O₃P
MDL Number:MFCD00009078
:228.23 g/mol
Boiling Pt:106…108 °C (1 mmHg)
Density:1.09 g/cm³
Flash Pt:>112 °C
EINECS:214-097-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.4980 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together