Order Entry
Czech Republic
ContactUsLinkComponent
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%
Irritant
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%
Certificates
undefined
Irritant
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%

Specification Test Results

  • Appearance:
    Light yellow to beige to pink Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    121 to 125 °C
  • Assay:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together