N,N-Diphenylcarbamoyl chloride, Sigma-Aldrich®
Catalog # SIAL153591-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:231.68 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:201-450-2
UN:3261
CAS Number:83-01-2
Formula:C₁₃H₁₀ClNO
MDL Number:MFCD00000633