L(-)-Methioninol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2899-37-8
:
Formula:C₅H₁₃NOS
MDL Number:MFCD00004735
:135.2 g/mol
Melting Pt:31…33 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:220-788-1
Related Information
Frequently Bought Together