Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl acetoacetate-[1,3-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Irritant
Ethyl acetoacetate-[1,3-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL485640-500MG
Supplier:  Merck
CAS Number:  
Irritant
Ethyl acetoacetate-[1,3-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL485640-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
 :GHS07
Storage Temperature:Ambient
CAS Number:77504-73-5
Formula:C₆H₁₀O₃