Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Home
Ethyl acetoacetate-[1,2,3,4-13C4]
Ethyl acetoacetate-[1,2,3,4-¹³C₄] (99% ¹³C ), Sigma-Aldrich®
Ethyl acetoacetate-[1,2,3,4-¹³C₄] (99% ¹³C ), Sigma-Aldrich®
SIAL489263-500MG
:
Merck
:
Ethyl acetoacetate-[1,2,3,4-¹³C₄] (99% ¹³C ), Sigma-Aldrich®
SIAL489263-500MG
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
500 mg
Seq:
2725
:
:
M-Clarity™ quality level = MQ200
:
GHS07
:
Ambient
:
84508-55-4
:
C₆H₁₀O₃