1-(Trifluoroacetyl)imidazole, Sigma-Aldrich®
Catalog # SIAL346128-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:164.09 g/mol
EINECS:216-282-5
CAS Number:1546-79-8
Formula:C₅H₃F₃N₂O
MDL Number:MFCD00014501