O,O-Diethyl methylphosphonothionate, Sigma-Aldrich®
Catalog # SIAL443360-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
CAS Number:6996-81-2
Formula:C₅H₁₃O₂PS