
(Methyl)triphenoxyphosphonium iodide 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17579-99-6
:
Formula:C₁₉H₁₈IO₃P
MDL Number:MFCD00011911
Melting Pt:141…146 °C
EINECS:241-551-9
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Orange to brown Moist powder or crystals
- Infrared spectrum:Conforms
- Titration Argentometric:≥94 %
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together