Ethephon(2-chloroethylphosphonic acid), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16672-87-0
:
Formula:C₂H₆ClO₃P
MDL Number:MFCD00002142
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:240-718-3
UN:2928
Frequently Bought Together