
Diethyl (bromodifluoromethyl)phosphonate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65094-22-6
:
Formula:C₅H₁₀BrF₂O₃P
MDL Number:MFCD00069118
:267.01 g/mol
Boiling Pt:99…102 °C (16 mmHg)
Density:1.503
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together