
2-Chlorocyclohexanone 95% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:822-87-7
:
Formula:C₆H₉ClO
MDL Number:MFCD00001626
Boiling Pt:82…83 °C (10 mmHg)
Melting Pt:23 °C
Density:1.16 g/cm³
Flash Pt:82 °C
Storage Temperature:Refrigerator
EINECS:212-505-5
ADR:9,
:132.59 g/mol
:GHS07
Specification Test Results
- Appearance:Clear brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4830 to 1.4850 (20°C, 589 nm)
- GC:≥94 %
- Stabilizer:0.2 % CaCO3/MgO (1:1)
Specification Test Results
Frequently Bought Together