Order Entry
Czech Republic
ContactUsLinkComponent
1,1,1-Trifluoro-2-butanone, Sigma-Aldrich®
1,1,1-Trifluoro-2-butanone, Sigma-Aldrich®
Catalog # SIAL542628-5G
Supplier:  Merck
CAS Number:  
1,1,1-Trifluoro-2-butanone, Sigma-Aldrich®
Catalog # SIAL542628-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :126.08 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:381-88-4
Formula:C₄H₅F₃O
MDL Number:MFCD00190641