Order Entry
Czech Republic
ContactUsLinkComponent
7-Methoxy-2-naphthol, Sigma-Aldrich®
7-Methoxy-2-naphthol, Sigma-Aldrich®
Catalog # SIAL367508-5G
Supplier:  Merck
CAS Number:  
7-Methoxy-2-naphthol, Sigma-Aldrich®
Catalog # SIAL367508-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :174.2000 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:5060-82-2
Formula:C₁₁H₁₀O₂
MDL Number:MFCD00075494