
Perfluorobutane-1-sulphonyl chloride ≥98%
Catalog # H27437.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:318.55 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
CAS Number:2991-84-6
Formula:C₄ClF₉O₂S
MDL Number:MFCD03412262