
4-Fluorobenzenesulphonamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:402-46-0
:
Formula:C₆H₆FNO₂S
MDL Number:MFCD00025384
Melting Pt:124…127 °C
Storage Temperature:Ambient
EINECS:206-946-2
UN:2811
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together