ContactUsLinkComponent
1-Hexanesulphonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Irritant
1-Hexanesulphonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Certificates
undefined
Irritant
1-Hexanesulphonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Certificates
Specification Test Results
  • Appearance:
    White Crystalline solid
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 % (150°C)
  • UV:
    (0.5 M in water)
  • Titration after Ion exchange:
    ≥99.0 % (on dry substance)
  • Membrane filtration test:
    (on 0.5 M soln.) Passes test
  • Suitability:
    (HPLC gradient) Passes test