Order Entry
Czech Republic
ContactUsLinkComponent
Trifluoroacetic acid ≥99.9% for peptide synthesis
Irritant
Corrosive
Trifluoroacetic acid ≥99.9% for peptide synthesis
Catalog # 84578.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
Trifluoroacetic acid ≥99.9% for peptide synthesis
Catalog # 84578.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.9 %
  • Appearance:
    Clear colourless liquid
  • Density (20/4):
    1.475 - 1.500
  • Identification:
    Passes test
  • Residue on evaporation:
    Max. 0.002 %
  • Water:
    Max. 0.02 %
  • Absorbance (260 nm):
    Max. 1
  • Absorbance (300 nm):
    Max. 0.03

Specification Test Results

About this item

 :
 :114.02 g/mol
Merck Index:13,09754
Boiling Pt:71.8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.490 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
 :GHS07
 :GHS05
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169