Order Entry
Czech Republic
ContactUsLinkComponent
SE 260 Mighty Small II Deluxe Mini Vertical Electrophoresis Unit
SE 260 Mighty Small II Deluxe Mini Vertical Electrophoresis Unit
Certifikáty
undefined
SE 260 Mighty Small II Deluxe Mini Vertical Electrophoresis Unit

Specifikace

  • Catalog No:
  • 732-2598
  • 732-1765
  • Model:
  • SE 260
  • SE 260
  • Glass plate size (W×H):
  • 100×80 mm
  • 100×80 mm
  • Humidity:
  • Up to 80%
  • Up to 80%
  • Max. temperature:
  • 45 °C
  • 45 °C
  • Spacers:
  • 0,75 mm
  • 0,75 mm
  • Unit W×H×D:
  • 165×180×160 mm
  • 165×180×160 mm
  • Weight:
  • 1,50 kg

Specifikace

Combs

Informace o produktu

Frequently Bought Together