Water, SupraSolv® for headspace gas chromatography, Supelco®
: Merck
:7732-18-5
:
:H₂O
:MFCD00011332
:18.02 g/mol
:13,10098
:100 °C (1013 hPa)
:0 °C
:1 g/cm³ (4 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:231-791-2
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Molar Mass:18.02 g/mol
- Boiling Point:100 °C (1013 hPa)
- Vapor Pressure:23 hPa (20 °C)
- Density:1.00 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
Frequently Bought Together