Order Entry
Czech Republic
ContactUsLinkComponent
Cleanroom wipes, BioClean Oryx™
Cleanroom wipes, BioClean Oryx™
Supplier:  Nitritex
Certificates
undefined
Cleanroom wipes, BioClean Oryx™
Supplier:  Nitritex
Certificates

Specifications

  • Catalog No:
  • NITRBOWS-12
  • NITRBOWS-9B
  • NITRBOWS-9
  • NITRBOWB-9
  • NITRBOWB-12
  • Model:
  • BioClean Oryx™
  • BioClean Oryx™
  • BioClean Oryx™
  • BioClean Oryx™
  • BioClean Oryx™
  • Basis weight:
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • Absorbency - extrinsic:
  • 382 ml/m²
  • 382 ml/m²
  • 382 ml/m²
  • 382 ml/m²
  • 382 ml/m²
  • Absorbency - intrinsic:
  • 5,6 ml/g
  • 5,6 ml/g
  • 5,6 ml/g
  • 5,6 ml/g
  • 5,6 ml/g
  • Ions - calcium:
  • 0,18 ppm
  • 0,18 ppm
  • 0,18 ppm
  • 0,18 ppm
  • 0,18 ppm
  • Ions - sodium:
  • 0,47 ppm
  • 0,47 ppm
  • 0,47 ppm
  • 0,47 ppm
  • 0,47 ppm

Specifications

Related Information

Frequently Bought Together