ContactUsLinkComponent
Adapters, receiver, vacuum, bent
Adapters, receiver, vacuum, bent
Catalog # WITG1.906.025
Supplier:  witeg Labortechnik
undefined
Adapters, receiver, vacuum, bent
Catalog # WITG1.906.025
Supplier:  witeg Labortechnik
Specifications
  • Cone size:
    29/32 NS
  • Socket size:
    19/26 NS
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3,3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Receiver adapters , bent (angle: 105°), with NS socket and NS cone, vacuum connection: glass olive, with drain tip.
Certifications: DIN 12594