ContactUsLinkComponent
Column head with valve stopcock acc. to Antlinger
Column head with valve stopcock acc. to Antlinger
Catalog # WITG1.610.029
Supplier:  witeg Labortechnik
undefined
Column head with valve stopcock acc. to Antlinger
Catalog # WITG1.610.029
Supplier:  witeg Labortechnik
Specifications
  • Cone size:
    29/32 NS
  • Pk:
    1
Specifications
About this item
Borosilicate glass3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Columns heads acc. to Antlinger, with PTFE-fine-dosing valve, cone to cloumn NS 29/32.