Orders LinkContactUsLinkComponent
Swab in tubes
Swab in tubes
Supplier:   LP ITALIANA
Swab in tubes
Supplier:  LP ITALIANA
Certificates
3 Options Available Below
Product Details & Documents
Sterile dry swabs in tubes.
Head is made of rayon or cotton.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Shaft/head
    Sterile
    Ø
    Length
    Packed
    Availability
    Price
  • LPIT111298
    Swabs in PS tubes
    Swabs in PS tubes
    Plastic/cotton
    +
    16 mm
    150 mm
    Individually
    Log in for availability
    Log in for availability
    1 * 200 KS
    1 * 200 KS - 6 310,00 Kč
    6 310,00 Kč
    1 * 200 KS
       6 310,00 Kč
  • LPIT111598
    Swabs in PP tube
    Swabs in PP tube
    Plastic/rayon
    +
    12 mm
    150 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 KS
    1 * 1.000 KS - 18 680,00 Kč
    18 680,00 Kč
    1 * 1.000 KS
       18 680,00 Kč
  • LPIT111698
    Swabs in PP tubes
    Swabs in PP tubes
    Plastic/rayon
    +
    12 mm
    150 mm
    Individually
    Log in for availability
    Log in for availability
    1 * 500 KS
    1 * 500 KS - 13 510,00 Kč
    13 510,00 Kč
    1 * 500 KS
       13 510,00 Kč
This product has been blocked by your organization. Please contact your purchasing department for more information