ContactUsLinkComponent
Buttres, reaction vessels components, DURAN®
Buttres, reaction vessels components, DURAN®
Catalog # SCOT210335705
Supplier:  DWK Life Sciences
undefined
Buttres, reaction vessels components, DURAN®
Catalog # SCOT210335705
Supplier:  DWK Life Sciences
Specifications
  • Ext.Ø:
    184 mm
  • With:
    flat flange DN 150
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • For flat flanges DN 150
Buttress, end ground without groove.