
1-Allyl-3-methylimidazolium chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65039-10-3
:
Formula:C₇H₁₁ClN₂
MDL Number:MFCD07784450
:158.63 g/mol
Melting Pt:49…51 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together