Organic reference standard, Inosine 10 µg/ml in methanol
Catalog # PROONE9279
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:58-63-9
- Description:Inosine 10 µg/ml in methanol
- Formula:C₁₀H₁₂N₄O₅
- Description:Inosine