Order Entry
Czech Republic
ContactUsLinkComponent
 
Octyl-β-D-thioglucopyranoside 98%
 
undefined
Octyl-β-D-thioglucopyranoside 98%

 

  • Appearance:
    White Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 % (ELSD)
  • Specific optical rotation:
    -52° to -54° (20°C, 589 nm) (c=1, CH3OH)

 

 

Frequently Bought Together