Order Entry
Czech Republic
ContactUsLinkComponent
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 100 mmol/l in aqueous solution, Sigma-Aldrich®
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 100 mmol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALC8552-25UMO
Supplier:  Merck
CAS Number:  
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 100 mmol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALC8552-25UMO
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 µmol
  • Seq:
    9125

Specifications

About this item

 :
 :527.12 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:252-849-3
CAS Number:36051-68-0
Formula:C₉H₁₄N₃Na₂O₁₄P₃
MDL Number:MFCD00078193