Pepstatin A
Katalogové číslo 786-052
Dodavatel: G-Biosciences
Číslo CAS:
Specifikace
- Pk:25 mg
- CAS No.:26305-03-3
- Storage conditions:Refrigerator
- MDL No.:MFCD00060740
- Protein/peptide name:Pepstatin A
- Molecular formula:C₃₄H₆₃N₅O₉
Specifikace
O tomto produktu
Pepstatin is a potent inhibitor of various aspartic proteases, including cathepsin D, renin, pepsin, bacterial aspartic proteases and HIV proteases. Soluble in DMSO or methanol.