Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:36051-68-0
:
Formula:C₉H₁₄N₃Na₂O₁₄P₃
MDL Number:MFCD00078193
:527.12 g/mol
EINECS:252-849-3
Frequently Bought Together