Order Entry
Czech Republic
ContactUsLinkComponent
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Catalog # SIALA1852-1VL
Supplier:  Merck
CAS Number:  
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Catalog # SIALA1852-1VL
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Seq:
    0032

Specifications

About this item

 :
 :551.14 g/mol (anhy)
EINECS:213-579-1
CAS Number:34369-07-8
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MDL Number:MFCD00150755