Order Entry
Czech Republic
ContactUsLinkComponent
Adenosine-5'-triphosphate disodium salt (ATP disodium salt)
Adenosine-5'-triphosphate disodium salt (ATP disodium salt)
Catalog # A1348.0100
Supplier:  PanReac AppliChem
CAS Number:  
Adenosine-5'-triphosphate disodium salt (ATP disodium salt)
Catalog # A1348.0100
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :551.14 g/mol
EINECS:213-579-1
CAS Number:987-65-5
 :MFCD00080339
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃