
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate, Ultrapure
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:16178-48-6
:
Formula:C₁₀H₁₃N₅Na₂O₁₀P₂·2H₂O
MDL Number:MFCD00066635
:471.167 g/mol (anhy)
EINECS:240-314-7
:GHS07
Storage Temperature:Freezer
Specification Test Results
- A250/A260:0.74 – 0.82
- A280/A260:0.13 – 0.19
- Arsenic:<0.0001%
- ATP:≤1%
- Em (259 nm, Phosphate Buffer, pH 7.0):≥14500
- Heavy Metals:<0.002%
- Loss on Drying:<8%
- Solubility (5%, Water):pass
Specification Test Results
Frequently Bought Together