Order Entry
Czech Republic
ContactUsLinkComponent
 
 
2'-Deoxyadenosine monohydrate 99%
 
undefined
 
2'-Deoxyadenosine monohydrate 99%

 

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤8 % (K.F.)
  • Sulfated ash:
    ≤0.1 % (800°C)
  • HPLC:
    ≥98.5 %
  • Appearance of solution:
    (c=1, w/v, 1M NaOH) clear colorless to almost colorless
  • Heavy metals (as Pb):
    ≤10 ppm

 

 

Frequently Bought Together