Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloroadenosine hemihydrate, Sigma-Aldrich®
2-Chloroadenosine hemihydrate, Sigma-Aldrich®
Catalog # SIAL861863-50MG
Supplier:  Merck
CAS Number:  
2-Chloroadenosine hemihydrate, Sigma-Aldrich®
Catalog # SIAL861863-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :621.3880 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:205-678-3
CAS Number:81012-94-4
Formula:C₁₀H₁₂ClN₅O₄·0.5H₂O
MDL Number:MFCD00149351