Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2',5'-Dideoxyadenosine
2',5'-Dideoxyadenosine, Sigma-Aldrich®
2',5'-Dideoxyadenosine, Sigma-Aldrich®
SIALD7408-5MG
:
Merck
:
2',5'-Dideoxyadenosine, Sigma-Aldrich®
SIALD7408-5MG
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 mg
Seq:
2210
:
:
235.2425 g/mol
:
M-Clarity™ quality level = MQ100
:
6698-26-6
:
C₁₀H₁₃N₅O₂
:
MFCD00210904