
Naphthol AS-MX phosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1596-56-1
:
Formula:C₁₉H₁₈NO₅P
MDL Number:MFCD00042711
:M-Clarity™ quality level = MQ200
EINECS:216-480-1
:GHS07
Storage Temperature:Freezer
Frequently Bought Together