Order Entry
Czech Republic
ContactUsLinkComponent
1,2-Bis(dichlorophosphino)benzene, SAFC®
1,2-Bis(dichlorophosphino)benzene, SAFC®
Catalog # SIAL698490-250MG
Supplier:  Merck
CAS Number:  
1,2-Bis(dichlorophosphino)benzene, SAFC®
Catalog # SIAL698490-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
CAS Number:82495-67-8
Formula:C₆H₄Cl₄P₂