Order Entry
Czech Republic
ContactUsLinkComponent
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) for synthesis, Sigma-Aldrich®
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    650 g/l (20 °C)
  • Melting Point:
    192 - 194 °C
  • Molar Mass:
    148.11 g/mol
  • Bulk Density:
    800 kg/m3
  • pH:
    7.0 - 11.0 (20 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together