Order Entry
Czech Republic
ContactUsLinkComponent
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) 98%
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) 98%
Certificates
undefined
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) 98%

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    191 to 195 °C
  • Titration after Ion exchange:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together